Search
Search
| 產品號碼 | Description | For Use With (Equipment) |
|---|---|---|
| 6036.2520 | 2-Position/6-Port (1500 bar) | Vanquish Column Compartment H |
| 6036.2530 | 6-Position/7-Port (1500 bar) | Vanquish Column Compartment H |
| 6230.1520 | 2-Position/6-Port (700 bar) | Vanquish Column Compartment C |
| 6230.1530 | 6-Position/7-Port (700 bar) | Vanquish Column Compartment C |
| 6250.1520 | Low-Dispersion 2-Position/6-Port (1500 bar) | Vanquish Column Compartment N |
Thermo Scientific™ Vanquish™ Column Compartments support up to two column switching valves. Thermo Scientific™ Vanquish™ Horizon and Vanquish™ Core Switching Valves are used in Vanquish Column Compartments H and C, respectively, for UHPLC and HPLC applications. Vanquish Low-Dispersion Switching Valves have an optimized design for use with Vanquish Column Compartment N for all nano-, capillary-, and micro-flow LC and LC-MS applications.