Viper™ TQ Capillary Kits
Thermo Scientific™
Viper™ TQ Capillary Kits
Boost your productivity and confidence, and accelerate method development using application-specific LC workflows.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)
6036.2807AAutomated Method Scouting
6036.2010AInverse Gradient for standard- free quantitation with charged aerosol detection
6036.2020ATandem LC and LC-MS
6036.2030ATrap Heart-Cut and Simple Switch 2D-LC
Catalog number 6036.2807A
Price (KRW)
-
For Use With (Application):
Automated Method Scouting
Improve chromatographic resolution by minimizing dead volume with the Thermo Scientific™ Viper™ TQ UHPLC Fittings. These pre-assembled kits allow you to quickly connect LC modules, valves, and columns for leak-free fluidic HPLC and UHPLC connections. Ideally suited for running application-specific workflows with your HPLC and LC-MS systems, like the Thermo Scientific™ Vanquish™ Method Development LC Systems, Thermo Scientific Vanquish Online 2D-LC Systems, or Thermo Scientific Vanquish Duo UHPLC Systems.
Enjoy the confidence of robust and reliable fingertight LC connections giving you tactile and audible feedback. Tailored to your advanced workflow requirements, Viper TQ application-specific capillary kits offer:
  • Simplified fluidic connection for all user levels with torque-controlled fingertight connections with sensory feedback
  • Easy-to-install, tool-free reusable capillaries requiring up to 40% less force compared to non-torque controlled fittings
  • Reliable, reproducible LC connections that deliver low carryover and simplify method transfer
Specifications
For Use With (Application)Automated Method Scouting
MaterialMP35N
Max. Pressure1517 bar
For Use With (Equipment)Vanquish Horizon and Flex UHPLC systems
Product LineViper TQ
TypeFor Vanquish Automatic Method Scouting Systems
Unit SizeEach