Vanquish™ HPLC and UHPLC System Accessories
Thermo Scientific™
Vanquish™ HPLC and UHPLC System Accessories
Maximize your liquid chromatography (LC) performance with system accessories best suited for the Thermo Scientific™ Vanquish™ HPLC and UHPLC systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesFor Use With (Application)Product Line
6036.9003Rear Seal and Needle Wash Tubing Labels for Vanquish SystemsLabeling rear seal and needle wash tubing in Vanquish UHPLC SystemsVanquish HPLC and UHPLC Systems
6261.1130Viper Guard ConnectorVanquish Binary Pump N or Metering Device, Viper TQ in Viper only ValvesViper, nano Viper, Viper TQ
6268.0130PEEK Mesh HPLC Solvent FiltersEluent filteringVanquish HPLC and UHPLC Systems
Catalog number 6036.9003
Price (MXN)
-
Includes:
Rear Seal and Needle Wash Tubing Labels for Vanquish Systems
For Use With (Application):
Labeling rear seal and needle wash tubing in Vanquish UHPLC Systems
Product Line:
Vanquish HPLC and UHPLC Systems
Extend the compatibility and lifespan of nanoViper™ Fignertight Fitting Systems and Viper™ TQ UHPLC Fittings with the Viper Guard Connector. Add the Thermo Scientific™ PEEK Mesh HPLC solvent filter and Rear Seal and Needle Wash Tubing Labels for Vanquish LC Systems for enhance user-friendliness.
Optimizing the chromatographic performance and user-friendliness with accessories suited best for Vanquish HPLC and UHPLC systems.
Specifications
For Use With (Application)Labeling rear seal and needle wash tubing in Vanquish UHPLC Systems
IncludesRear Seal and Needle Wash Tubing Labels for Vanquish Systems
For Use With (Equipment)Vanquish Horizon, Flex UHPLC and Core HPLC Systems
Product LineVanquish HPLC and UHPLC Systems
TypeLabel
Unit SizeEach