Search
| Número de catálogo | Para utilizar con (aplicación) | Incluye |
|---|---|---|
| 6036.9010 | Accessory Kit | Reconditioner, union and wrench |
| 6261.5020 | Reconditioning of Viper TQ fittings | Viper TQ reconditioner |
| 6261.5060 | Connecting Viper TQ capillaries and Viper TQ capillaries with (nano)Viper capillaries | Viper TQ Union, Titan |
| 6261.5065A | Column Compatibility | Viper TQ Adapter, SST (2 pcs.) |
| 6212.0003 | Fluidic connection for LC systems | Viper TQ Tee piece, Titanium |
We offer the following accessories for Viper TQ UHPLC Fittings: