CYL, Pump 10 CC CLEANED 140C

Catalog number: 603843

Applied Biosystems™  Related applications: Protein Sequencing & Peptide Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is a replacement part for the 140C Instrument.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Procise Protein Sequencer
Product Line: Applied Biosystems™
Product Size: 1 unit
Shipping Condition: Room Temperature
Type: Pump

Contents & storage

Store at room temperature.

Documents

Manuals & protocols