Keep your Thermo Scientific™ Dionex™ UltiMate™ 3000 System free of water condensation by using this kit.
Have Questions?
Catalog Number
For Use With (Equipment)
6040.0005
Ultimate 3000 Systems
Catalog number 6040.0005
Price (EUR)
-
For Use With (Equipment):
Ultimate 3000 Systems
Keep your system free of water condensation with the Thermo Scientific™ System Drain Kit for Dionex™ UltiMate™ 3000 Systems. The kit includes all components required for system drainage and detailed installation instructions.