Conventional stainless steel HPLC connectors use a ferrule and a nut to establish a connection. A flaw of this design is that a void volume-free connection is not guaranteed, particularly when changing the tubing between differently shaped threads (e.g. when changing a column).
Viper connectors are standard for Dionex RSLC system connections
Compatible with virtually any third party UHPLC or HPLC instrument, valve, and column hardware
規格
DescriptionExtension Kit for Automated Method Scouting
Diameter (English) Outer1/8 in.
Styleextension kit to increase solvent/buffer choices
For Use With (Equipment)UltiMate 3000 Quaternary SD/RS AMS Setup