Product Image
Thermo Scientific™
UltiMate™ 3000 Pump Piston Seals and Support Rings
Order replacement piston seals and support rings for your Thermo Scientific™ Dionex™ UltiMate™ 3000.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
6040.0306Dionex UltimMate 3000 DS Pump, Dionex UltiMate 3000 RS Pump, Dionex UltiMate 3000 BM PumpPiston Seal (in Plate of Seal Wash System) NP
6266.0305Dionex UltiMate 3000 RS PumpPiston Seal (Main Piston Seal) RP
6266.0304Dionex UltiMate 3000 SD PumpPiston Seal (Main Piston Seal) RP
6040.9011Dionex UltiMate 3000 BX PumpPiston Seal Kit (Main Piston Seal) NP
6040.9010Dionex UltiMate 3000 BX PumpPiston Seal Kit (Main Piston Seal) RP
6040.0012Dionex UltiMate 3000 SD Pump, Dionex UltiMate 3000 RS PumpSupport Rings for Piston Seals
6025.2012Dionex UltiMate 3000 BM PumpPiston Seal Kit (Main Piston Seal)
6043.0295Dionex UltiMate 3000 XRS PumpPiston Seal Kit (Main Piston Seal) RP
Catalog number 6040.0306
Price (MXN)
-
For Use With (Equipment):
Dionex UltimMate 3000 DS Pump, Dionex UltiMate 3000 RS Pump, Dionex UltiMate 3000 BM Pump
Type:
Piston Seal (in Plate of Seal Wash System) NP
Order Thermo Scientific™ Dionex™ UltiMate™ 3000 Piston Seals and Support Rings as replacements when needed. Available individually or as kits, the seals and support rings for UltiMate 3000 pumps are specific for either normal-phase or reversed-phase chromatography. Kits include either support rings or secondary seals.
Specifications
For Use With (Equipment)Dionex UltimMate 3000 DS Pump, Dionex UltiMate 3000 RS Pump, Dionex UltiMate 3000 BM Pump
TypePiston Seal (in Plate of Seal Wash System) NP
Unit SizeEach