Maintenance Kits for UltiMate™ 3000 Pumps
Actual product may vary
Thermo Scientific™

Maintenance Kits for UltiMate™ 3000 Pumps

Maintenance Kits for UltiMate™ 3000 Pumps vary by pump.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
6040.1952DGP-3600 SD Pump
6040.1950ISO-3100 SD Pump
6040.1951LPG-3400 SD Pump
6040.1953HPG-3 x 00 SD Pump
6040.1954ALPG-3400 RS Pump
6040.1955ADGP-3600 RS Pump
6040.1956AHPG-3 x 00 RS Pump
6042.1950ISO-3100 BM Pump
6042.1951LPG-3400 BM Pump
6042.1952DGP-3600 BM Pump
6042.1953LPG-3400 BX (RP) Pump
6042.1954LPG-3400 BX (NP) Pump
6043.1954LPG-3400 XRS Pump
Catalog number 6040.1952
Price (CLP)
-
For Use With (Equipment):
DGP-3600 SD Pump

Thermo Scientific™ Dionex Maintenance Kits for UltiMate™ 3000 Pumps vary by pump.

Each maintenance kit includes:

  • Solvent line filter
  • Silicone tubing
  • PharMed tubing
  • Tubing connectors
  • RP piston seals
  • Support rings
  • O-ring seal
  • Valve cartridge
  • Cap seal for purge valve knob
  • Cleaning swabs
Specifications
DescriptionMaintenance Kit for DGP-3600 SD pump
For Use With (Equipment)DGP-3600 SD Pump
TypeMaintenance Kit
Unit SizeEach