Viper™ and nanoViper™ Fingertight Fittings Accessories
Actual product may vary
Viper™ and nanoViper™ Fingertight Fittings Accessories
Viper™ and nanoViper™ Fingertight Fittings Accessories
Thermo Scientific™
Viper™ and nanoViper™ Fingertight Fittings Accessories
Obtain reproducible chromatographic results using tool-free and easy-to-use Thermo Scientific™ Viper™ and Thermo Scientific™ nanoViper™ Fingertight Fittings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
6040.2303Plug
6040.2304Union
6040.2312Calibration Tool
6036.1045Filter
Catalog number 6040.2303
Price (MXN)
-
Type:
Plug
Ensure optimal separations and maximum LC performance using virtually zero-dead-volume connections with the Thermo Scientific™ Viper™ Fingertight Fitting systems. Accessories help complete the installation according to your application needs.
See Viper and nanoViper resources
We offer the following accessories for Viper Fingertight Fittings Systems:
  • Union exclusively designed for connecting Viper and nanoViper Fingertight Fittings directly without additional dead volume
  • Viper Plug for installation in open valve positions to protect the bores or for diagnostic functions
  • Tool to recalibrate Viper and nanoViper fittings for an extended lifetime when connected to multiple columns with non-standard column bore geometry
  • Viper Inline Filter (titanium, av. frit pre size 0.5μm) to protect the column from contaminants
  • Adaptor for Viper Fingertight Fittings (2 pcs, stainless steel, for columns ≥ 2.1 mm ID) to adapt to non-standard column bore geometry
Ma
Specifications
DescriptionViper Plug
For Use With (Application)Covering open ports, diagnostic
MaterialTitanium
Max. Pressure1500 bar
For Use With (Equipment)All HPLC and UHPLC Systems
Product LineViper
TypePlug
Unit SizeEach