Search
| Catalog Number | For Use With (Equipment) | Diameter (Metric) Outer |
|---|---|---|
| 6040.2308 | HPG Pumps for RS Systems | 130 μm/180 μm |
| 6040.2301 | ISO, LPG, or DGP Pumps for RS Systems | 130 μm/180 μm |
| 6040.2302 | ISO, LPG, or DGP Pumps for SD Systems | 180 μm |
| 6040.2309 | HPG Pumps for SD Systems | 180 μm |
Avoid data-destroying dead volumes in the capillary connections of your HPLC or UHPLC pump connections by using Thermo Scientific™ Viper™ Capillary Kits for LC Pumps. This kit provides worry-free connections for Thermo Scientific™ Dionex™ UltiMate™ 3000 ISO, LPG, or DGP Pumps in UltiMate RSLC and SD systems. Even with conventional columns and moderate pressures, poor connections can cause retention time shifts and peak distortions. The kit contains ViperFingertight Fittings which seal at the tip of the tubing and eliminate dead volumes and make it easier to plumb your HPLC or UHPLC system.
You won’t have to worry about capillary slippage or improper ferrule positioning anymore. Consistently perfect connections ensure robust chromatography with minimal interruption.
Pump-Specific KitsStainless-steel Viper capillaries (I.D. x L, one each):