Viper™ Online SPE Solutions Kits
Viper™ Online SPE Solutions Kits
Actual product may vary
Thermo Scientific™
Viper™ Online SPE Solutions Kits
Ensure a perfect connection between your online SPE and your HPLC/UHPLC system with a single kit with zero-dead-volume fittings and capillaries.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
6040.2801Dionex Ultimate 3000 x2 Dual RS System
6040.2802Dionex UltiMate 3000 x2 Dual SD System
Catalog number 6040.2801
Price (MXN)
-
For Use With (Equipment):
Dionex Ultimate 3000 x2 Dual RS System
Ensure a perfect connection between your online SPE and your HPLC/UHPLC system with the Thermo Scientific™ Dionex™ Viper™ Online SPE Solution Kits. A single kit contains Viper zero-dead-volume fittings and capillaries you need to work with your x2 Dual system. The kit contains Viper Fingertight Fittings that seal at the tip of the tubing and eliminate dead volumes and make it easier to plumb your HPLC or UHPLC system.
See Viper and nanoViper resources
Conventional stainless steel HPLC connectors use a ferrule and a nut to establish a connection. A flaw of this design is that a void volume-free connection is not guaranteed, particularly when changing the tubing between differently shaped threads (e.g. when changing a column).
  • Viper connectors are standard for Dionex RSLC system connections
  • Compatible with virtually any third party UHPLC or HPLC instrument, valve, and column hardware
Specifications
Diameter (Metric) Outer180 μm
For Use With (Application)Online SPE for Matrix Elimination and Diluted Samples
Length (Metric)Various
MaterialStainless Steel
Max. Pressure18125 psi (1250 bar)
QuantityEach
For Use With (Equipment)Dionex Ultimate 3000 x2 Dual RS System
Product LineViper
TypeOnline SPE Solutions Kit
Unit SizeEach