Search
| Catalog Number | For Use With (Equipment) | Diameter (Metric) Outer |
|---|---|---|
| 6040.2803 | Dionex Ultimate 3000 x2 Dual RS System | 130 μm |
| 6040.2804 | Dionex UltiMate 3000 x2 Dual SD System | 180 μm |
Order the Thermo Scientific™ Dionex™ Viper™ Tandem Operation Capillary Kit for all the connectors and capillaries you need to plumb your tandem LC system. This single kit contains Viper Fingertight Fittings technology, which avoids data-destroying dead volumes in the capillary connections of your tandem HPLC or UHPLC system. Even with conventional columns and moderate pressures, poor connections can cause retention time shifts and peak distortions. Viper Fingertight Fittings seal at the tip of the tubing to eliminate dead volumes and make it easier to plumb your HPLC or UHPLC system.