Viper™ Parallel Setup Solutions Kits
Actual product may vary
Viper™ Parallel Setup Solutions Kits
Viper™ Parallel Setup Solutions Kits
Viper™ Parallel Setup Solutions Kits
Thermo Scientific™
Viper™ Parallel Setup Solutions Kits
Order this single kit for all the easy, zero-dead-volume fittings and capillaries for your parallel LC system.
Have Questions?
카탈로그 번호용도 (장비)
6040.2810Dionex UltiMate 3000 x2 Dual SD System
카탈로그 번호 6040.2810
제품 가격(KRW)
-
용도 (장비):
Dionex UltiMate 3000 x2 Dual SD System
Obtain all the connectors and capillaries you need to plumb parallel systems and ensure consitently perfect connections. The Thermo Scientific™ Dionex™ Viper™ Parallel Setup Solutions Kit contains Viper Fingertight Fittings which avoid data-destroying dead volumes in capillary connections of your parallel HPLC or UHPLC system. Even with conventional columns and moderate pressures, poor connections can cause retention time shifts and peak distortions. Viper Fingertight Fittings seal at the tip of the tubing to eliminate dead volumes and make it easier to plumb your HPLC or UHPLC system.
See Viper and nanoViper resources
Conventional stainless steel HPLC connectors use a ferrule and a nut to establish a connection. A flaw of this design is that a void volume-free connection is not guaranteed, particularly when changing the tubing between differently shaped threads (e.g. when changing a column).
  • Viper connectors are standard for Dionex RSLC system connections
  • Compatible with virtually any third party UHPLC or HPLC instrument, valve, and column hardware
사양
직경(미터법) 외부180 μm
길이(미터법)Various
물질Stainless Steel
최대 압력18125 psi (1250 bar)
수량Each
용도 (장비)Dionex UltiMate 3000 x2 Dual SD System
유형Parallel LC Operation
Unit SizeEach