Search
| Catalog Number | For Use With (Equipment) | Diameter (Metric) Outer |
|---|---|---|
| 6040.2819 | Dionex UltiMate 3000 x2 Dual SD System | 180 μm |
| 6040.2820 | Dionex Ultimate 3000 x2 Dual RS System | 130 μm/180 μm |
Avoid data-destroying dead volumes in the capillary connections of your inverse gradient HPLC or UHPLC system. The Thermo Scientific™ Dionex™ Viper™ Inverse Gradient Kit contains Viper Fingertight Fittings which avoid data-destroying dead volumes in capillary connections and ensure consitently perfect connections. Even with conventional columns and moderate pressures, poor connections can cause retention time shifts and peak distortions. Viper Fingertight Fittings seal at the tip of the tubing to eliminate dead volumes and make it easier to plumb your HPLC or UHPLC system.