Viper™ Inverse Gradient Kits
Actual product may vary
Thermo Scientific™
Viper™ Inverse Gradient Kits
Order this single kit for all the easy, zero-dead-volume fittings and capillaries for your inverse gradient LC system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Diameter (Metric) Outer
6040.2820Dionex Ultimate 3000 x2 Dual RS System130 μm/180 μm
6040.2819Dionex UltiMate 3000 x2 Dual SD System180 μm
Catalog number 6040.2820
Price (EUR)
-
For Use With (Equipment):
Dionex Ultimate 3000 x2 Dual RS System
Diameter (Metric) Outer:
130 μm/180 μm
Avoid data-destroying dead volumes in the capillary connections of your inverse gradient HPLC or UHPLC system. The Thermo Scientific™ Dionex™ Viper™ Inverse Gradient Kit contains Viper Fingertight Fittings which avoid data-destroying dead volumes in capillary connections and ensure consitently perfect connections. Even with conventional columns and moderate pressures, poor connections can cause retention time shifts and peak distortions. Viper Fingertight Fittings seal at the tip of the tubing to eliminate dead volumes and make it easier to plumb your HPLC or UHPLC system.
See Viper and nanoViper resources
Specifications
DescriptionDual Gradient Operation
Diameter (Metric) Outer130 μm/180 μm
For Use With (Application)HPLC, UHPLC
Length (Metric)Various
MaterialStainless Steel
Max. Pressure18125 psi (1250 bar)
QuantityEach
For Use With (Equipment)Dionex Ultimate 3000 x2 Dual RS System
Product LineViper
TypeInverse Gradient Kit
Unit SizeEach