Thermo Scientific™
nanoViper™ Fingertight Fittings, 150µm
 Related applications: Industrial Chromatography
Maximize nano liquid chromatography (LC) performance and convenience with Thermo Scientific™ nanoViper™ Fingertight Fittings. Virtually zero-dead-volume connections ensure optimal separation efficiencies. Preassembled and finger-tightened, nanoViper fingertight fittings enable easy, tool-free instrument setup and configuration, and prevent overtightening. High pressure tested, these fittings provide UHPLC compatibility up to 1200 bar (17,400 psi).
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Robust nano LC Performance
  • Ferrule-free tightening design for virtually zero-dead-volume connections at ultrahigh pressures
  • Corrects alignment to avoid under- and overtightening
  • Suitable for temperatures up to 120°C
Easy to Use
  • Fingertight design
  • 1/32 in. PEEK sheathed fused silica
  • No tools required
  • Eliminates assembly of PEEK sleeve fittings
Flexible
  • Compatible with virtually any valve and any column from any manufacturer
  • Available individually or as complete application kits
Recommended Use
  • Use with Thermo Scientific™ UltiMate™ 3000 RSLCnano systems for fully integrated workflows
  • nanoViper connections are standard on UltiMate 3000 RSLCnano accessories and columns
Documents
Manuals & protocols