Order Thermo Scientific™ Dionex™ UltiMate™ 3000 RSLC Nano System Flow Meters when you need replacements. They are available for nano- capillary- and micro-LC flow rates.
Have Questions?
Change view
Catalog Number
For Use With (Equipment)
6041.7902A
Capillary LC (0.5 to 10 μL/Min)
6041.7901A
Nano LC (50 to 1000 nL/Min)
6041.7903A
Micro LC (5 to 50 μL/Min)
3 Options
Catalog number 6041.7902A
Price (MXN)
-
For Use With (Equipment):
Capillary LC (0.5 to 10 μL/Min)
Order Thermo Scientific™ Dionex™ UltiMate™ 3000 RSLC Nano System Flow Meters when you need replacements. They are available for nano- capillary- and microbore-LC flow rates.
Specifications
Biocompatible VersionYes
For Use With (Equipment)Capillary LC (0.5 to 10 μL/Min)