UltiMate™ 3000 RSLC Nano System Flow Meters
Product Image
Thermo Scientific™
UltiMate™ 3000 RSLC Nano System Flow Meters
Order Thermo Scientific™ Dionex™ UltiMate™ 3000 RSLC Nano System Flow Meters when you need replacements. They are available for nano- capillary- and micro-LC flow rates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
6041.7903AMicro LC (5 to 50 μL/Min)
6041.7901ANano LC (50 to 1000 nL/Min)
6041.7902ACapillary LC (0.5 to 10 μL/Min)
Catalog number 6041.7903A
Price (MXN)
-
For Use With (Equipment):
Micro LC (5 to 50 μL/Min)
Order Thermo Scientific™ Dionex™ UltiMate™ 3000 RSLC Nano System Flow Meters when you need replacements. They are available for nano- capillary- and microbore-LC flow rates.
Specifications
Biocompatible VersionYes
For Use With (Equipment)Micro LC (5 to 50 μL/Min)
TypeFlow Meter, Biocompatible with Flow Selector
Unit SizeEach