Search Thermo Fisher Scientific
Maximize your liquid chromatography (LC) performance with the Thermo Scientific™ Viper™ Fingertight Fitting system with biocompatible MP35N capillaries.
Catalog Number | Description |
---|---|
6042.2315 | Viper MP35N, 0.180 mm x 150 mm |
6042.2308 | Viper MP35N 0.180 x 65 mm |
6042.2322 | Viper MP35N 0.180 x 300 mm |
6042.2327 | Viper MP35N, 0.180 mm x 250 mm |
6042.2337 | Viper MP35N, 0.180 mm x 350 mm |
6042.2365 | Viper MP35N, 0.180 mm x 450 mm |
6042.2355 | Viper MP35N, 0.180 mm x 550 mm |
6042.2380 | Viper MP35N, 0.180 mm x 650 mm |
6042.2375 | Viper MP35N, 0.180 mm x 750 mm |
6042.2385 | Viper MP35N, 0.180 mm x 950 mm |
Maximize liquid chromatography (LC) performance and ensure optimal separations with virtually zero-dead-volume connections. Thermo Scientific™ Viper™ Fingertight Fitting Systems with biocompatible MP35N capillaries are easy to use and enable a fast, tool-free fluidic setup of biocompatible HPLC and UHPLC systems. These systems are compatible with virtually any column and valve type and suitable for fluidic connections with pressures up to 1517 bar (22,000 psi).
Robust LC Performance
Easy to Install
Flexible
Recommended Use
Thermo Scientific™ Vanquish™ Flex and Vanquish Horizon UHPLC systems and Thermo Scientifc™ UltiMate™ 3000 BioRS systems are equipped with the Viper Fingertight Fitting System with MP35N capillaries as standard.