Thermo Scientific™
Viper™ Capillary MP35N Fingertight Fittings, 130µm
Related applications:
Industrial Chromatography
Maximize liquid chromatography (LC) performance and get virtually zero-dead-volume connections to ensure optimal separations. Thermo Scientific™ Viper™ Capillary MP35N Fingertight Fittings for HPLC and UHPLC Systems are biocompatible, iron-free, easy to use and enable fast, tool-free setup, even for the most advanced column- and valve-switching configurations. Compatible with virtually any common column and valve type, it is suitable for system pressures of up to 1500 bar (22,000 psi). This 130μm I.D. biocompatible fitting is available in 750mm length.
Back to top
Robust LC Performance
  • Ferrule-free tightening design for virtually zero-dead-volume connections at operating pressures of up to 1500 bar
  • Suitable for temperatures up to 120°C
Easy to Use
  • Fingertight design
  • Flexible biocompatible MP35N capillaries (1/32 in o.d.)
  • No tools required
Flexible
  • Compatible with virtually any valve and any column from any manufacturer
  • Fits narrow connections such as 10-port valves
  • Enables mixed use with different designs
  • Available in different inner diameters: 100µm (0.004 in.), 130µm (0.005 in.) or 180µm (0.007 in.)
Recommended Use
Use with Thermo Scientific™ Vanquish™ LC systems, Thermo Scientific™ UltiMate™ 3000 LC systems, or any LC system for fully integrated workflows. Vanquish and UltiMate LC systems are equipped with the Viper fingertight fitting system as a standard.