Thermo Scientific™
Viper™ Capillary Kit for UltiMate™ 3000WPS or OAS Autosamplers
Related applications:
Industrial Chromatography
Avoid data-destroying dead volumes in the capillary connections of your HPLC or UHPLC autosampler by using the Thermo Scientific™ Dionex™ Viper™ Capillary Kit for UltiMate™ 3000 WPS and OAS autosamplers. Viper Fingertight Fittings seal at the tip of the tubing to eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system. A single kit contains all the connectors and capillaries you need and ensures consistently perfect connections.
Back to top
Description
Avoid data-destroying dead volumes in the capillary connections of your HPLC or UHPLC autosampler by using the Thermo Scientific™ Dionex™ Viper™ Capillary Kit for UltiMate™ 3000 WPS and OAS autosamplers. Viper Fingertight Fittings seal at the tip of the tubing to eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system. A single kit contains all the connectors and capillaries you need and ensures consistently perfect connections.