Transcend™ TurboFlow Systems
Thermo Scientific™
Transcend™ TurboFlow Systems
Perform high throughput UHPLC sample preparation, analysis, and clean-up for LC-MS with multichannel UHPLC systems.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoModelLC Channels
60500-60301-02Transcend VTLX-1 System1
60500-60202Transcend TLX-2 System2
60500-60203Transcend TLX-4 System4
Número do catálogo 60500-60301-02
Preço (BRL)
-
Model:
Transcend VTLX-1 System
LC Channels:
1
Enable fast cleanup of complex matrices for accelerated online sample preparation, lower your laboratory costs, and improve data quality using Thermo Scientific Transcend TLX Systems with TurboFlow technology. These systems use a combination of chromatographic techniques for online sample cleanup to improve LC-MS detection, reduce manual work, and provide higher productivity while the systems' additional channel capabilities improve throughput by up to 400%.
  • Transcend VTLX-1: Single-channel LC-MS with online sample cleanup
  • Transcend TLX-2: Two-channel LC-MS with online sample cleanup
  • Transcend TLX-4: Four-channel LC-MS with online sample cleanup
View Transcend System resourcesDiscover VanquishHPLC Support Center
Transcend TLX systems, which feature TurboFlow technology, combine diffusion, chemistry, and size exclusion to perform selective online sample cleanup of dirty or difficult matrices prior to HPLC or UHPLC separation and MS analysis. TurboFlow technology minimizes manual sample preparation and reduces overall sample preparation time by up to 95%.
Other benefits of TurboFlow technology include:
  • Minimization of manual sample preparation and facilitation of direct sample injection into an LC-MS system, including plasma, urine, food, and other complex sample matrices.
  • Reduction of ion suppression via higher specificity.
  • Reduction of work time through simplification of complex sample preparation protocols.
  • Simplified method development by allowing the same method to be used for different matrices.
Aria MX software optimizes performance and productivity by controlling pump operation, valve switching, cleaning, and gradient procedures on the Transcend system. The system is compatible with XCalibur or TraceFinder mass spectrometry software, with an intuitive graphical user interface that makes it simple to perform TurboFlow methods.
Especificações
AutosamplerVanquish Dual Split Sampler FT
LC Channels1
ModelTranscend VTLX-1 System
Pump1 Vanquish Quaternary Pump F + 1 Vanquish Binary Pump F
Unit SizeEach