SOLA™ SPE Plates
Actual product may vary
SOLA™ SPE Plates
SOLA™ SPE Plates
SOLA™ SPE Plates
Thermo Scientific™
SOLA™ SPE Plates
Innovative Thermo Scientific™ SOLA™ Solid Phase Extraction Plates deliver clean, highly reproducible sample extracts with lower elution volumes—for greater sensitivity, reliability and cost savings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionSorbent
60509-00130 mg/2 mLHRP (Reversed Phase Polymeric)
60309-00110 mg/2 mLHRP (Reversed Phase Polymeric)
60309-00210 mg/2 mLMixed Mode Strong Cation Exchanger, Polymeric
60509-00230 mg/2 mLMixed Mode Strong Cation Exchanger, Polymeric
60309-00310 mg/2 mLMixed Mode Strong Anion Exchanger, Polymeric
60509-00330 mg/2 mLMixed Mode Strong Anion Exchanger, Polymeric
60309-00410 mg/2 mLMixed Mode Weak Cation Exchanger, Polymeric
60509-00430 mg/2 mLMixed Mode Weak Cation Exchanger, Polymeric
60309-00510 mg/2 mLMixed Mode Weak Anion Exchanger, Polymeric
60509-00530 mg/2 mLMixed Mode Weak Anion Exchanger, Polymeric
Catalog number 60509-001
Price (BRL)
-
Description:
30 mg/2 mL
Sorbent:
HRP (Reversed Phase Polymeric)
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ SOLA™ Solid Phase Extraction (SPE) Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, SOLA SPE Plates deliver unparalleled performance, including:
  • Higher levels of reproducibility
  • Lower elution volumes
  • High extract cleanliness
SOLA SPE Plates achieve excellent recovery levels even with low volumes of extract solvents, resulting in a more concentrated analyte and increased sensitivity. You’ll also realize additional cost and time saving from faster sample dry-down time and reduced solvent usage.
  • 10mg/2mL 96 well plate format
  • Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX) and Strong Anion Exchanger (SAX) phase types
  • SOLA products are also available in cartridge format.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description30 mg/2 mL
For Use With (Application)Bioanalytical Applications - Hydrophobic Retention of Neutral Compounds with Complementary Retention of Polar Analytes. An All-Purpose Phase.
Bed Weight30 mg
Product LineSOLA
SorbentHRP (Reversed Phase Polymeric)
Type96-well SPE Plate
Volume (Metric)2 mL
Unit SizeEach