UltiMate™ 3000 Variable Wavelength Detector Flow Cells
Actual product may vary
UltiMate™ 3000 Variable Wavelength Detector Flow Cells
UltiMate™ 3000 Variable Wavelength Detector Flow Cells
UltiMate™ 3000 Variable Wavelength Detector Flow Cells
Thermo Scientific™
UltiMate™ 3000 Variable Wavelength Detector Flow Cells
Choose the Thermo Scientific™ Dionex™ UltiMate™ 3000 Variable Wavelength Detector Flow Cell based on the flow rate you need.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeMaterial
6074.0270Nano Flow CellFused Silica
6074.0320Semi-Preparative Flow CellPEEK
6074.0200Analytical Flow CellPEEK
6074.0250Analytical Flow CellStainless Steel
6074.0280Capillary Flow CellFused Silica
6074.0285Ultra-Low Dispersion UV-MonitorFused Silica
6074.0290Mic Flow CellFused Silica
6074.0300Semi-Micro Flow CellPEEK
6074.0360
also known as 6074.036
Semi-Micro Flow CellStainless Steel
Catalog number 6074.0270
Price (EUR)
-
Type:
Nano Flow Cell
Material:
Fused Silica
Choose the Thermo Scientific™ Dionex™ UltiMate™ 3000 Variable Wavelength Detector Flow Cell based on the flow rate you need. They are available in a variety of diameters for analytical, semi-analytical, semi-preparative and semi-micro flow rates.
Specifications
Diameter (Metric) Outer10 mm
For Use With (Application)Nano LC
MaterialFused Silica
Max. Pressure2900 psi (200 bar)
Volume (Metric)3 nL
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
Product LineUltiMate
TypeNano Flow Cell
Unit SizeEach