Thermo Scientific™
UltiMate™ 3000 Variable Wavelength Detector Flow Cells
Catalog number: 6074.0300
Thermo Scientific™
UltiMate™ 3000 Variable Wavelength Detector Flow Cells
Catalog number: 6074.0300
Choose the Thermo Scientific™ Dionex™ UltiMate™ 3000 Variable Wavelength Detector Flow Cell based on the flow rate you need. They are available in a variety of diameters for analytical, semi-analytical, semi-preparative and semi-micro flow rates.
Catalog Number
6074.0300
Unit Size
Each
For Use With (Equipment)
UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
Type
Semi-Micro Flow Cell
Material
PEEK
Diameter (Metric) Outer
7 mm
Price (USD)
Full specifications
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialPEEK
Max. Pressure725 psi (50 bar)
Product LineUltiMate
Volume (Metric)2.5 µL
TypeSemi-Micro Flow Cell
Diameter (Metric) Outer7 mm
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
Dionex UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
Type
Semi-Preparative Flow Cell
Material
PEEK
Diameter (Metric) Outer
0.4 mm
Price (USD)
Full specifications
For Use With (Application)Micro-LC
For Use With (Equipment)Dionex UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialPEEK
Max. Pressure1450 psi (100 bar)
Product LineUltiMate
Volume (Metric)0.7 µL
TypeSemi-Preparative Flow Cell
Diameter (Metric) Outer0.4 mm
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
Type
Analytical Flow Cell
Material
PEEK
Diameter (Metric) Outer
10 mm
Price (USD)
Full specifications
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialPEEK
Product LineUltiMate
Volume (Metric)11 µL
TypeAnalytical Flow Cell
Diameter (Metric) Outer10 mm
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
Type
Analytical Flow Cell
Material
Stainless Steel
Diameter (Metric) Outer
10 mm
Price (USD)
Full specifications
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialStainless Steel
Product LineUltiMate
Volume (Metric)11 µL
TypeAnalytical Flow Cell
Diameter (Metric) Outer10 mm
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
Type
UZ-View Flow Cell
Material
Fused Silica
Diameter (Metric) Outer
10 mm
Price (USD)
Full specifications
For Use With (Application)Nano LC
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialFused Silica
Max. Pressure2900 psi (200 bar)
Product LineUltiMate
Volume (Metric)3 nL
TypeUZ-View Flow Cell
Diameter (Metric) Outer10 mm
Unit SizeEach
Catalog Number
6074.0280
also known as 6074.028
Unit Size
Each
For Use With (Equipment)
UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
Type
UZ-View Flow Cell
Material
Fused Silica
Diameter (Metric) Outer
10 mm
Price (USD)
Full specifications
For Use With (Application)Capillary LC
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialFused Silica
Max. Pressure2900 psi (200 bar)
Product LineUltiMate
Volume (Metric)45 nL
TypeUZ-View Flow Cell
Diameter (Metric) Outer10 mm
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
Dionex UltiMate 3000 Variable Wavelength Detector
Type
Ultra-Low Dispersion UV-Monitor
Material
Fused Silica
Diameter (Metric) Outer
Price (USD)
Full specifications
For Use With (Application)Micro-LC
For Use With (Equipment)Dionex UltiMate 3000 Variable Wavelength Detector
MaterialFused Silica
Product LineUltiMate
Volume (Metric)45 nL
TypeUltra-Low Dispersion UV-Monitor
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
Dionex UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
Type
UZ-View Flow Cell
Material
Fused Silica
Diameter (Metric) Outer
10 mm
Price (USD)
Full specifications
For Use With (Application)Micro-LC
For Use With (Equipment)Dionex UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialFused Silica
Max. Pressure2900 psi (200 bar)
Product LineUltiMate
Volume (Metric)180 nL
TypeUZ-View Flow Cell
Diameter (Metric) Outer10 mm
Unit SizeEach
Catalog Number
6074.0360
also known as 6074.036
Unit Size
Each
For Use With (Equipment)
UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
Type
Semi-Micro Flow Cell
Material
Stainless Steel
Diameter (Metric) Outer
7 mm
Price (USD)
Full specifications
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialStainless Steel
Max. Pressure1740 psi (120 bar)
Product LineUltiMate
Volume (Metric)2.5 µL
TypeSemi-Micro Flow Cell
Diameter (Metric) Outer7 mm
Unit SizeEach
Showing 9 of 9
Catalog NumberSpecificationsUnit SizeFor Use With (Equipment)TypeMaterialDiameter (Metric) OuterPrice (USD)
6074.0300Full specifications
EachUltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Semi-Micro Flow CellPEEK7 mmRequest A Quote
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialPEEK
Max. Pressure725 psi (50 bar)
Product LineUltiMate
Volume (Metric)2.5 µL
TypeSemi-Micro Flow Cell
Diameter (Metric) Outer7 mm
Unit SizeEach
6074.0320Full specifications
EachDionex UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Semi-Preparative Flow CellPEEK0.4 mmRequest A Quote
For Use With (Application)Micro-LC
For Use With (Equipment)Dionex UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialPEEK
Max. Pressure1450 psi (100 bar)
Product LineUltiMate
Volume (Metric)0.7 µL
TypeSemi-Preparative Flow Cell
Diameter (Metric) Outer0.4 mm
Unit SizeEach
6074.0200Full specifications
EachUltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Analytical Flow CellPEEK10 mmRequest A Quote
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialPEEK
Product LineUltiMate
Volume (Metric)11 µL
TypeAnalytical Flow Cell
Diameter (Metric) Outer10 mm
Unit SizeEach
6074.0250Full specifications
EachUltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Analytical Flow CellStainless Steel10 mmRequest A Quote
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialStainless Steel
Product LineUltiMate
Volume (Metric)11 µL
TypeAnalytical Flow Cell
Diameter (Metric) Outer10 mm
Unit SizeEach
6074.0270Full specifications
EachUltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)UZ-View Flow CellFused Silica10 mmRequest A Quote
For Use With (Application)Nano LC
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialFused Silica
Max. Pressure2900 psi (200 bar)
Product LineUltiMate
Volume (Metric)3 nL
TypeUZ-View Flow Cell
Diameter (Metric) Outer10 mm
Unit SizeEach
6074.0280
also known as 6074.028
Full specifications
EachUltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)UZ-View Flow CellFused Silica10 mmRequest A Quote
For Use With (Application)Capillary LC
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialFused Silica
Max. Pressure2900 psi (200 bar)
Product LineUltiMate
Volume (Metric)45 nL
TypeUZ-View Flow Cell
Diameter (Metric) Outer10 mm
Unit SizeEach
6074.0285Full specifications
EachDionex UltiMate 3000 Variable Wavelength DetectorUltra-Low Dispersion UV-MonitorFused SilicaRequest A Quote
For Use With (Application)Micro-LC
For Use With (Equipment)Dionex UltiMate 3000 Variable Wavelength Detector
MaterialFused Silica
Product LineUltiMate
Volume (Metric)45 nL
TypeUltra-Low Dispersion UV-Monitor
Unit SizeEach
6074.0290Full specifications
EachDionex UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)UZ-View Flow CellFused Silica10 mmRequest A Quote
For Use With (Application)Micro-LC
For Use With (Equipment)Dionex UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialFused Silica
Max. Pressure2900 psi (200 bar)
Product LineUltiMate
Volume (Metric)180 nL
TypeUZ-View Flow Cell
Diameter (Metric) Outer10 mm
Unit SizeEach
6074.0360
also known as 6074.036
Full specifications
EachUltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)Semi-Micro Flow CellStainless Steel7 mmRequest A Quote
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
MaterialStainless Steel
Max. Pressure1740 psi (120 bar)
Product LineUltiMate
Volume (Metric)2.5 µL
TypeSemi-Micro Flow Cell
Diameter (Metric) Outer7 mm
Unit SizeEach
Showing 9 of 9
Choose the Thermo Scientific™ Dionex™ UltiMate™ 3000 Variable Wavelength Detector Flow Cell based on the flow rate you need. They are available in a variety of diameters for analytical, semi-analytical, semi-preparative and semi-micro flow rates.