UltiMate™ 3000 Variable Wavelength Detector Flow Cells
Actual product may vary
UltiMate™ 3000 Variable Wavelength Detector Flow Cells
UltiMate™ 3000 Variable Wavelength Detector Flow Cells
UltiMate™ 3000 Variable Wavelength Detector Flow Cells
Thermo Scientific™
UltiMate™ 3000 Variable Wavelength Detector Flow Cells
Choose the Thermo Scientific™ Dionex™ UltiMate™ 3000 Variable Wavelength Detector Flow Cell based on the flow rate you need.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeMaterial
6074.0320Semi-Preparative Flow CellPEEK
6074.0200Analytical Flow CellPEEK
6074.0250Analytical Flow CellStainless Steel
6074.0270Nano Flow CellFused Silica
6074.0280Capillary Flow CellFused Silica
6074.0285Ultra-Low Dispersion UV-MonitorFused Silica
6074.0290Mic Flow CellFused Silica
6074.0300Semi-Micro Flow CellPEEK
6074.0360
also known as 6074.036
Semi-Micro Flow CellStainless Steel
Catalog number 6074.0320
Price (MXN)
-
Type:
Semi-Preparative Flow Cell
Material:
PEEK
Choose the Thermo Scientific™ Dionex™ UltiMate™ 3000 Variable Wavelength Detector Flow Cell based on the flow rate you need. They are available in a variety of diameters for analytical, semi-analytical, semi-preparative and semi-micro flow rates.
Specifications
Diameter (Metric) Outer0.4 mm
For Use With (Application)Micro-LC
MaterialPEEK
Max. Pressure1450 psi (100 bar)
Volume (Metric)0.7 μL
For Use With (Equipment)UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS)
Product LineUltiMate
TypeSemi-Preparative Flow Cell
Unit SizeEach