HPLC UV Detector Lamps
HPLC UV Detector Lamps
Actual product may vary
Thermo Scientific™
HPLC UV Detector Lamps
HPLC UV Detector Lamps can be used with Variable Wavelength, Diode Array, and Multiple-Wavelength detectors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
6074.2000Tungsten Lamp
6074.1110Deuterium Lamp
Catalog number 6074.2000
Price (BRL)
-
Type:
Tungsten Lamp
Choose Thermo Scientific™ HPLC UV Detector Lamps for use with Variable Wavelength, Diode Array, and Multiple-Wavelength detectors.
Specifications
Detection ModeUV Detection
For Use With (Equipment)DAD-3000(RS), MWD-3000(RS) and VWD-3x00(RS) Series Detector
TypeTungsten Lamp
Unit SizeEach