Thermo Scientific™
Vanquish™ Variable Wavelength Detector F Flow Cells
 Related applications:
Industrial Chromatography
Choose the Thermo Scientific™ Vanquish™ Variable Wavelength Detector F Flow Cell based on the flow rate needed. Flow cells are available for both analytical, micro, and semi-preparative flow rates. Choose the best flow cell based on your needs for the Thermo Scientific™ Vanquish™ Variable Wavelength Detector F.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Choose the Thermo Scientific™ Vanquish™ Variable Wavelength Detector F Flow Cell based on the flow rate needed. Flow cells are available for both analytical, micro, and semi-preparative flow rates. Choose the best flow cell based on your needs for the Thermo Scientific™ Vanquish™ Variable Wavelength Detector F.
Compatible with: Vanquish Variable Wavelength Detector F
Documents
Manuals & protocols