Choose a flow cell for your Dionex™ UltiMate™ 3000 Fluorescence Detector based on flow rate.
Have Questions?
Alterar visualização
Número do catálogo
Sample Capacity
6078.4330
2 μL
6078.4230
8 μL
2 opções
Número do catálogo 6078.4330
Preço (BRL)
-
Sample Capacity:
2 μL
Choose the Thermo Scientific™ Dionex™ UltiMate™ 3000 Fluorescence Detector Flow Cell based on the flow rate needed. Flow cells are available for both analytical and micro flow rates.
Especificações
For Use With (Equipment)Dionex Ultimate; 3000 Fluorescence Detector