Vanquish™ Fluorescence Detector F Flow Cells
Vanquish™ Fluorescence  Detector F Flow Cells
Actual product may vary
Thermo Scientific™
Vanquish™ Fluorescence Detector F Flow Cells
Choose the Vanquish Fluorescence Detector F Flow Cell based on the flow rate needed. Flow cells are available for both analytical and micro flow rates.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호부피(미터법)
6079.42308 μL
6079.43302 μL
카탈로그 번호 6079.4230
제품 가격(KRW)
-
부피(미터법):
8 μL
Choose the Thermo Scientific™ Vanquish™ Fluorescence Detector F Flow Cell based on the flow rate needed. Flow cells are available for both analytical and microflow rates. Choose the standard flow cell or the micro flow cell based on your needs for the Thermo Scientific™ Vanquish™ Fluorescence Detector F or as a biocompatible flow cell for the Thermo Scientific™ UltiMate™ 3000 Fluorescence Detector.
사양
설명Vanquish Fluorescence Standard Flow Cell
물질Biocompatible
부피(미터법)8 μL
용도 (장비)Vanquish Fluorescence Detector F or UltiMate 3000 Fluorescence Detector
제품라인Vanquish
유형Standard Fluorescence Flow Cell
Unit SizeEach