Thermo Scientific™
Analog Signal Output Kit for Dionex™ Corona™ Veo™ Detectors
 Related applications:
Industrial Chromatography
Use the Thermo Scientific™ Analog Signal Output Kit to obtain 0V to 1V output to the recording device or external A/D converter of your Thermo Scientific™ Dionex™ Corona™ Veo™ Detector.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Use the Thermo Scientific™ Analog Signal Output Kit to obtain 0V to 1V output to the recording device or external A/D converter of your Thermo Scientific™ Dionex™ Corona™ Veo™ Detector.
Documents
Manuals & protocols