Thermo Scientific™
UltiMate™ 3000 Diode Array Detector Flow Cells
Related applications:
Industrial Chromatography
Choose the Thermo Scientific™ UltiMate™ 3000 Diode Array Detector Flow Cell with the desired flow rate. Flow cells are available in analytical, semi-prep and semi-micro flow rates.
Back to top
Flow cells are available in both stainless steel and PEEK to meet your application requirements.