Capture more information from your sample by using this kit to upgrade your Thermo Scientific™ Dionex™ UltiMate™ MWD-3000RS detector to a DAD 3000RS detector.
Have Questions?
變更視圖
產品號碼
Type
For Use With (Equipment)
6082.3045
Upgrade Kit (MWD-3000RS to DAD-3000RS)
Dionex Ultimate MWD-3000 RS
6082.3035
Upgrade Kit (MWD-3000 to DAD-3000)
Dionex Ultimate MWD-3000
2 選項
產品號碼 6082.3045
價格 (HKD)
-
Type:
Upgrade Kit (MWD-3000RS to DAD-3000RS)
For Use With (Equipment):
Dionex Ultimate MWD-3000 RS
Capture more information from samples analyzed by LC. Use the Thermo Scientific™ Dionex™ UltiMate™ MWD-3000RS to DAD-3000RS Upgrade kit to upgrade from the multiwavelength detector to the diode array detector. This kit includes all components for upgrading an UltiMate MWD-3000 to an UltiMate DAD-3000 detector. The kit must be installed by authorized service personnel.
規格
Detection ModeUV-vis Detection (8 Channels)
For Use With (Equipment)Dionex Ultimate MWD-3000 RS