Thermo Scientific™
Vanquish™ LightPipe™ Flow Cells for the Vanquish™ Diode Array Detector HL
Related applications:
Industrial & Applied Science
The completely new design of the Thermo Scientific™ Vanquish™ Diode Array Detector (DAD) HL incorporates Thermo Scientific™ LightPipe™ Flow Cells, which provide the best signal-to-noise (S/N) performance through the combination of lowest baseline noise, a very long light path, and minimum peak dispersion. The ultra-wide dynamic range is ideal for simultaneous detection of highly concentrated main compounds and impurities down to trace levels. Interfering refractive index (RI) and thermal effects are reduced to a minimum for less baseline drift and therefore more reliable peak integration.
Back to top
Great standards in flow cells for a wide range of applications
Standard flow cell:
  • Volumes: 2µL (illuminated), 0.8µL (dispersion)
  • Flow Cell Pressure Limit 6MPa (60bar, 870psi)
High sensitivity flow cell:
  • Volumes: 13µL (illuminated), 4.0µL (dispersion)
  • Flow Cell Pressure Limit 6MPa (60bar, 870psi)