Thermo Scientific™
Vanquish™ Flow Cells for Vanquish™ Diode Array Detector FG
 Related applications:
Industrial Chromatography
Choose from a wide portfolio of Thermo Scientific™ Vanquish™ Flow Cells for the Thermo Scientific™ Vanquish™ Diode Array Detector FG. There's a flow cell to match your application-related requirements including low-dispersion for UHPLC separations, high-pressure resistance for LC-MS or fractionation, and compatibility with the analysis of biomolecules.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Great flow cells for a wide range of applications
12 MPa (120 bar, 1740 psi) flow cell pressure limit:
  • Standard flow cell
  • Semi-analytical flow cell
  • Semi-micro flow cells (biocompatible and SST version available)
5 MPa (50 bar, 720 psi) flow cell pressure limit:
  • Standard biocompatible flow cell
Documents
Manuals & protocols