Thermo Scientific™
Laboratory Refrigerator and Freezer Casters
 Related applications:
Cold Storage
Achieve higher elevation on high-performance refrigerators and freezers with Thermo Scientific™ Laboratory Refrigerator and Freezer Casters. Casters offer a 2 or 3 in. height increase for Thermo Scientific™ Revco™ , Forma™ and Puffer Hubbard™ laboratory refrigerators and freezers.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • An option for replacement on most models
  • Factory-installed when ordered with unit; customer-installed if ordered separately
  • 2 in. (50mm) casters are standard on high-performance refrigerators and freezers
  • Optional 3 in. (76mm) casters are shipped with the cabinet but must be installed upon delivery
Ordering Information:
  • 6041: 2 in. casters (50mm), set of four
  • 6041-1: 3 in. casters (76mm), set of four
  • 6983: Locking casters for -30°C freezers only, includes two nonlocking 2 in. (50mm) casters
Documents
Manuals & protocols