Acetonitrile, for HPLC
Acetonitrile, for HPLC
Acetonitrile, for HPLC
Thermo Scientific Chemicals

Acetonitrile, for HPLC

CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Catalog NumberQuantityPackaging
6100100404 LEcoSafPak™
Catalog number 610010040
Price (USD)
424.65
Online Exclusive
472.00
Save 47.35 (10%)
Each
Quantity:
4 L
Packaging:
EcoSafPak™
Request bulk or custom format
Price (USD)
424.65
Online Exclusive
472.00
Save 47.35 (10%)
Each
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Refractive index1.3405 to 1.3425 (25°C, 589 nm)
Water=<0.01 % (K.F.)
Titrable base=<0.0006 meq/g
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only