Acetonitrile, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetonitrile, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Catalog number 610220010
Price (USD)
136.65
Online Exclusive
152.00
Save 15.35 (10%)
Each
Request bulk or custom format
Price (USD)
136.65
Online Exclusive
152.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Appearance (Form)Liquid
Assay>=99.9 %
Water=<50 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only