sec-Butanol, 99+%, Extra Dry, AcroSeal™
sec-Butanol, 99+%, Extra Dry, AcroSeal™
sec-Butanol, 99+%, Extra Dry, AcroSeal™
sec-Butanol, 99+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

sec-Butanol, 99+%, Extra Dry, AcroSeal™

CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 610261000
Price (EUR)
42,10
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
42,10
Each
Chemical Identifiers
CAS78-92-2
IUPAC Namebutan-2-ol
Molecular FormulaC4H10O
InChI KeyBTANRVKWQNVYAZ-UHFFFAOYNA-N
SMILESCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.0 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Water=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only