Thermo Scientific Chemicals

Pyridine, 99.0%, Extra Dry, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 610440010
Each
Thermo Scientific Chemicals

Pyridine, 99.0%, Extra Dry, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 610440010
Each
Catalog number: 610440010
Price (USD)
Price: 170.00
Your price:
Quantity
-

Chemical Identifiers

CAS
110-86-1
IUPAC Name
pyridine
Molecular Formula
C5H5N
InChI Key
JUJWROOIHBZHMG-UHFFFAOYSA-N
SMILES
C1=CC=NC=C1
Appearance (Color)
Colorless
Appearance (Form)
Liquid
Assay
>=99.0 %
Water
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text