Thermo Scientific™
Richard-Allan Scientific™ Signature Series™ Pen-Fix™
Related applications:
Anatomical Pathology
Achieve fixation and dehydration of specimens simultaneously with Thermo Scientific™ Richard-Allan Scientific™ Signature Series™ Pen-Fix™, a uniquely buffered alcoholic formalin fixative.
Back to top
  • Excellent frozen section fixative
  • Performs well with fatty or thick tissue sections
  • Penetration rate three times faster than 10% Neutral Buffered Formalin
  • Can be used on all open or enclosed tissue processors and in the grossing room
  • Recommended for use in the second and/or third stations of the processor as a secondary fixative