Methyl sulfoxide, 99.7%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Methyl sulfoxide, 99.7%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Methyl sulfoxide, 99.7%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Methyl sulfoxide, 99.7%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl sulfoxide, 99.7%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
610971000100 mL
Catalog number 610971000
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
93.20
Save 13.55 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
93.20
Save 13.55 (15%)
Each
Specifications
Assay Percent Range99.7%
Beilstein01, IV, 1277
CAS67-68-5
ChEBICHEBI:28262
Chemical Name or MaterialMethyl sulfoxide
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only