Deionized Water Support for Recirculating Chillers
Imagem ilustrativa. O produto pode variar
Thermo Scientific™
Deionized Water Support for Recirculating Chillers
Maintain the resistivity of your cooling water with Thermo Scientific™ Deionized Water Support for Recirculating Chillers.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoFor Use With (Equipment)
611006000006Merlin Chillers with a PD1/PD2 pump
611003000017System IV heat exchangers, 115 V
611006000009System III heat exchangers, 120 V
598000000010ThermoFlex with IDI
611003000016System III heat exchangers, 115 V
Número do catálogo 611006000006
Preço (BRL)
-
For Use With (Equipment):
Merlin Chillers with a PD1/PD2 pump
Maintain the resistivity of your cooling water with Thermo Scientific™ Deionized Water Packages.
  • Maintains a water resistivity level between 1 and 3 MOhmcm
Includes: Kits include a filter canister, DI media, plumbing, connectors, filter, and a LED indication to let you know when the media needs to be replaced. DI Cartridges include only the replacement DI media.
Especificações
For Use With (Equipment)Merlin Chillers with a PD1/PD2 pump
IncludesFilter canister, DI Media, plumbing, connectors, filter, and a LED indicator to let you know when the media needs to be replaced (requires 230V 50Hz power source)
Electrical Requirements230 V 50 Hz
TypeDeionized Water Support
Unit SizeEach