Karl Fischer reagent, stabilized, Thermo Scientific Chemicals
Karl Fischer reagent, stabilized, Thermo Scientific Chemicals
Karl Fischer reagent, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Karl Fischer reagent, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 611130010
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Specifications
CAS7553-56-2
ChEBICHEBI:17606
Chemical Name or MaterialKarl Fischer reagent
Density0.93g/mL
IUPAC Namediiodine
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only