Ammonium Citrate Dibasic, 98%, Thermo Scientific Chemicals
Ammonium Citrate Dibasic, 98%, Thermo Scientific Chemicals
Ammonium Citrate Dibasic, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium Citrate Dibasic, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Catalog number 611160025
Price (USD)
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS3012-65-5
IUPAC Namediammonium 3-carboxy-3-hydroxypentanedioate
Molecular FormulaC6H14N2O7
InChI KeyYXVFQADLFFNVDS-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Residue after ignition=<0.01 %
Chloride (Cl)=<0.001 %
Heavy metals=<5 ppm
Iron (Fe)=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only