Bismuth Subnitrate, 79%, Thermo Scientific Chemicals
Bismuth Subnitrate, 79%, Thermo Scientific Chemicals
Bismuth Subnitrate, 79%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth Subnitrate, 79%, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
611355000500 g
Catalog number 611355000
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS1304-85-4
IUPAC Namebis(nitrooxy)bismuthanyl nitrate
Molecular FormulaBiN3O9
InChI KeyPPNKDDZCLDMRHS-UHFFFAOYSA-N
SMILES[O-][N+](=O)O[Bi](O[N+]([O-])=O)O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
IdentificationPasses test
Assay>=79.0 % (as Bi2O3 dried basis)
Loss on drying=<3.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only