Thermo Scientific Chemicals

Cedarwood oil

Catalog number: 612085000
500 mL, EcoSafPak™, Each
Thermo Scientific Chemicals

Cedarwood oil

Catalog number: 612085000
500 mL, EcoSafPak™, Each
Quantity
Packaging

Chemical Identifiers

CAS
8000-27-9
Appearance (Form)
Liquid
Specific gravity
(25°C) 0.948 to 0.970
Refractive index
1.502 to 1.507 (20°C, 589 nm)
Optical density
=<0.29
Solubility
(in alcohol) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text