Gentamycin Sulfate
Gentamycin Sulfate
Gentamycin Sulfate
Thermo Scientific Chemicals

Gentamycin Sulfate

CAS: 1405-41-0 | C21H43N5O7·H2SO4
Have Questions?
Catalog NumberQuantity
6139800101 g
Catalog number 613980010
Price (USD)
137.65
Online Exclusive
153.00
Save 15.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
137.65
Online Exclusive
153.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS1405-41-0
IUPAC Name(2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,6S)-3-amino-6-(aminomethyl)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol; (2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,6S)-3-amino-6-[(1R)-1-(methylamino)ethyl]oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol; (2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,6S)-3-amino-6-[(1R)-1-aminoethyl]oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol; sulfuric acid
Molecular FormulaC60H125N15O25S
InChI KeyRDEIXVOBVLKYNT-HDZPSJEVSA-N
SMILESOS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](CN)CC[C@H]3N)[C@@H]2O)OC[C@]1(C)O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@@H](CC[C@H]3N)[C@@H](C)N)[C@@H]2O)OC[C@]1(C)O.CN[C@H](C)[C@@H]1CC[C@@H](N)[C@@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Identificationpasses test
Specific optical rotation+107° to +121° (20°C, 589 nm)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only