Nalgene™ Tubing Y-Type Connectors
Thermo Scientific™
Nalgene™ Tubing Y-Type Connectors
Maximize liquid flow with non-tapered hose barbed Y-Type Connectors, molded of polypropylene resin, free of animal derived components.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With
6152-03753/8 in.
6152-01251/8 in. I.D.
6152-0125PK1/8 in. I.D.
126152-01251/8 in. Tubing
6152-01873/16 in. I.D.
6152-0187PK3/16 in. I.D.
6152-02501/4 in. I.D.
6152-0250PK1/4 in. I.D.
6152-03125/16 in. I.D.
6152-0312PK5/16 in. I.D.
6152-0375PK3/8 in.
6152-05001/2 in. Tubing
6152-0500PK1/2 in. Tubing
Catalog number 6152-0375
Price (USD)
302.00
Case of 48
For Use With:
3/8 in.
Price (USD)
302.00
Case of 48
Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.
  • Polypropylene construction
  • Autoclavable
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionY-Type Connector
For Use With3/8 in.
Packaging Quantity12/Pk., 48/Cs.
AutoclavableAutoclavable
Diameter (English) Inner3/8 in.
Diameter (Metric) Inner9.52 mm
MaterialPolypropylene
Product TypeTubing Connector
Unit SizeCase of 48